accessibility menu, dialog, popup

UserName

CAS RN 261952-24-3

NH2OHOOFFF

CAS RN 261952-24-3

IUPAC Name: 2-amino-2-[4-(trifluoromethoxy)phenyl]acetic acid
Synonyms: amino[4-(trifluoromethoxy)phenyl]acetic acid
Molecular Weight (g/mol): 235.16
Molecular Formula: C9H8F3NO3
InChi Key: KBWQXEWTAXZMRX-UHFFFAOYNA-N
SMILES: NC(C(O)=O)C1=CC=C(OC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)