accessibility menu, dialog, popup

UserName

CAS RN 26040-62-0

H3COOCl

CAS RN 26040-62-0

IUPAC Name: methyl 7-chloroheptanoate
Synonyms: methyl 7-chloroheptanoate
Molecular Weight (g/mol): 178.66
Molecular Formula: C8H15ClO2
InChi Key: YKSFGXAJDCLJKE-UHFFFAOYSA-N
SMILES: COC(=O)CCCCCCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results