accessibility menu, dialog, popup

UserName

CAS RN 25472-44-0

H3COOHNOOCH3

CAS RN 25472-44-0

IUPAC Name: ethyl 2-(2-ethoxy-2-oxoethyl)-1H-pyrrole-3-carboxylate
Synonyms: ethyl 2-(2-ethoxy-2-oxoethyl)-1H-pyrrole-3-carboxylate
Molecular Weight (g/mol): 225.24
Molecular Formula: C11H15NO4
InChi Key: YVLGULDSGMYQPE-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=C(C=CN1)C(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results