CAS RN 2540-56-9
CAS RN 2540-56-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Rumenic Acid (~80%), TRC
CAS: 2540-56-9 Molecular Formula: C18 H32 O2 Molecular Weight (g/mol): 280.45 Synonym: 9,11-Octadecadienoic acid, (E,Z)- (8CI),(9Z,11E)-9,11-Octadecadienoic acid,(9Z,11E)-Octadecadienoic acid,9-cis,11-trans-Linoleic acid,9-cis,11-trans-Octadecadienoic acid,9Z,11E-Octadecadienoic acid,Bovinic acid,Rumenic acid,cis-9,trans-11 Conjugated linoleic acid,cis-9,trans-11-Octadecadienoic acid,trans-11-cis-9-Octadecadienoic acid,9,11-Octadecadienoic acid, (9Z,11E)-,Rumenic Acid IUPAC Name: (9Z,11E)-octadeca-9,11-dienoic acid SMILES: CCCCCC\C=C\C=C/CCCCCCCC(=O)O