accessibility menu, dialog, popup

UserName

CAS RN 2480-28-6

CH3HN(S)NH2NH2NOHO

CAS RN 2480-28-6

IUPAC Name: (2S)-5-[(diaminomethylidene)amino]-2-(methylamino)pentanoic acid
Synonyms: (2S)-5-[(diaminomethylidene)amino]-2-(methylamino)pentanoic acid
Molecular Weight (g/mol): 188.23
Molecular Formula: C7H16N4O2
InChi Key: NTWVQPHTOUKMDI-YFKPBYRVSA-N
SMILES: CN[C@@H](CCCN=C(N)N)C(O)=O
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)