accessibility menu, dialog, popup

UserName

CAS RN 2435-50-9

NNO

CAS RN 2435-50-9

IUPAC Name: pyrimidine-4-carbaldehyde
Synonyms: pyrimidine-4-carbaldehyde
Molecular Weight (g/mol): 108.1
Molecular Formula: C5H4N2O
InChi Key: OKULHRWWYCFJAB-UHFFFAOYSA-N
SMILES: O=CC1=NC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results