CAS RN 2410-29-9
CAS RN 2410-29-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,3-Dioleoyl-2-stearoylglycerol, TRC
CAS: 2410-29-9 Molecular Formula: C57H106O6 Molecular Weight (g/mol): 887.47 Synonym: (9Z)-9-Octadecenoic Acid 1,1'-[2-[(1-Oxooctadecyl)oxy]-1,3-propanediyl] Ester,(Z)-9-Octadecenoic Acid 1,1'-[2-[(1-Oxooctadecyl)oxy]-1,3-propanediyl] Ester,2-Stearo-1,3-diolein,2-Stearoyldiolein,Glyceryl 1,3-Dioleate-2-stearate,Triglyceride OSO,sn,Triglyceride OStO,sn SMILES: CCCCCCCCCCCCCCCCCC(=O)OC(COC(=O)CCCCCCC\C=C/CCCCCCCC)COC(=O)CCCCCCC\C=C/CCCCCCCC