CAS RN 2338-54-7
CAS RN 2338-54-7
IUPAC Name:
1-fluoro-4-methoxy-2-methylbenzene
Synonyms:
1-fluoro-4-methoxy-2-methylbenzene
Molecular Weight (g/mol):
140.16
Molecular Formula:
C8H9FO
InChi Key:
XZBXPBDJLUJLEU-UHFFFAOYSA-N
SMILES:
COC1=CC=C(F)C(C)=C1
1
–
1
of
1
results
1
4-Fluoro-3-methylanisole, 98%, Thermo Scientific™
CAS: 2338-54-7 Molecular Formula: C8H9FO Molecular Weight (g/mol): 140.157 MDL Number: MFCD00042293 InChI Key: XZBXPBDJLUJLEU-UHFFFAOYSA-N Synonym: 4-fluoro-3-methylanisole,2-fluoro-5-methoxytoluene,1-fluoro-4-methoxy-2-methyl-benzene,benzene, 1-fluoro-4-methoxy-2-methyl,pubchem2623,4-fluoro-3-methylani,4-fluoro-3-methylanisol,3-methyl-4-fluoroanisole,4-fluoro-3-methylanizole,acmc-1cr5e PubChem CID: 137546 IUPAC Name: 1-fluoro-4-methoxy-2-methylbenzene SMILES: CC1=C(C=CC(=C1)OC)F