accessibility menu, dialog, popup

UserName

CAS RN 23239-35-2

CH3(S)H2NOHO

CAS RN 23239-35-2

IUPAC Name: (2S)-2-amino-2-methyl-3-phenylpropanoic acid
Synonyms: (2S)-2-amino-2-methyl-3-phenylpropanoic acid
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: HYOWVAAEQCNGLE-JTQLQIEISA-N
SMILES: C[C@](N)(CC1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)