CAS RN 23169-33-7
CAS RN 23169-33-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
9-(2-Hydroxyethyl)guanine, TRC
CAS: 23169-33-7 Molecular Formula: C7 H9 N5 O2 Molecular Weight (g/mol): 195.18 Synonym: 6H-Purin-6-one, 2-amino-1,9-dihydro-9-(2-hydroxyethyl)-,Guanine, 9-(2-hydroxyethyl)- (8CI),2-Amino-1,9-dihydro-9-(2-hydroxyethyl)-6H-purin-6-one,9-(2-Hydroxyethyl)guanine,9-(2'-Hydroxyethyl)guanine,NSC 40668,32W88,GW 318209X,Ph Eur Aciclovir Impurity P,32W88 IUPAC Name: 2-amino-9-(2-hydroxyethyl)-1H-purin-6-one SMILES: NC1=Nc2c(ncn2CCO)C(=O)N1