CAS RN 2309668-15-1
CAS RN 2309668-15-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
AR-13324 M1 metabolite, MedChemExpress
MedChemExpress AR-13324 M1 metabolite is a hydrolysis metabolite of AR-13324 mesylate. AR-13324 is a inhibitor of ROCK kinase.