accessibility menu, dialog, popup

UserName

CAS RN 230618-41-4

NNBr

CAS RN 230618-41-4

IUPAC Name: 2-bromo-6-(pyrrolidin-1-yl)pyridine
Synonyms: 2-bromo-6-(pyrrolidin-1-yl)pyridine
Molecular Weight (g/mol): 227.11
Molecular Formula: C9H11BrN2
InChi Key: HZOWJDJNPISFJC-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=N1)N1CCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results