accessibility menu, dialog, popup

UserName

CAS RN 22699-70-3

CH3CH3O

CAS RN 22699-70-3

IUPAC Name: 1-(3-ethylphenyl)ethan-1-one
Synonyms: 3-ethylacetophenone
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: ZRYRILAFFDKOPB-UHFFFAOYSA-N
SMILES: CCC1=CC=CC(=C1)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results