CAS RN 2243-35-8
CAS RN 2243-35-8
IUPAC Name:
2-oxo-2-phenylethyl acetate
Synonyms:
2-oxo-2-phenylethyl acetate
Molecular Weight (g/mol):
178.19
Molecular Formula:
C10H10O3
InChi Key:
BGAXCPSNMHVHJC-UHFFFAOYSA-N
SMILES:
CC(=O)OCC(=O)C1=CC=CC=C1
1
–
2
of
2
results
1
2-(Acetyloxy)-1-phenylethanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-(Acetyloxy)-1-phenylethanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.