CAS RN 22155-91-5
CAS RN 22155-91-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
L-Pyroglutamic Acid Beta-Naphthylamide, TRC
CAS: 22155-91-5 Molecular Formula: C15 H14 N2 O2 Molecular Weight (g/mol): 254.28 Synonym: L-Pyroglutamic Acid -Ss-Naphthylamide IUPAC Name: (2S)-N-naphthalen-2-yl-5-oxopyrrolidine-2-carboxamide SMILES: O=C(Nc1ccc2ccccc2c1)[C@@H]3CCC(=O)N3