accessibility menu, dialog, popup

UserName

CAS RN 220465-43-0

HOBOHHN

CAS RN 220465-43-0

IUPAC Name: (1H-indol-4-yl)boronic acid
Synonyms: 1H-indol-4-ylboronic acid
Molecular Weight (g/mol): 160.97
Molecular Formula: C8H8BNO2
InChi Key: USJUQEVUEBCLLR-UHFFFAOYSA-N
SMILES: OB(O)C1=C2C=CNC2=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)