CAS RN 2190-30-9
CAS RN 2190-30-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Palmitodiolein, MedChemExpress
MedChemExpress Palmitodiolein (Triglyceride POO) is a triacylglycerol which is present in vegetable oils.