CAS RN 2163056-91-3
CAS RN 2163056-91-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
BI-1347, MedChemExpress
MedChemExpress BI-1347 is a potent CDK8 inhibitor extracted from patent WO2017202719A1, product I-003, has an IC50 of 1.1 nM.