accessibility menu, dialog, popup

UserName

CAS RN 216144-91-1

OHBHOOHO(E)

CAS RN 216144-91-1

IUPAC Name: (2E)-3-[3-(dihydroxyboranyl)phenyl]prop-2-enoic acid
Synonyms: (2E)-3-[3-(dihydroxyboranyl)phenyl]prop-2-enoic acid
Molecular Weight (g/mol): 191.98
Molecular Formula: C9H9BO4
InChi Key: QCHIEOGZUMAQKI-SNAWJCMRSA-N
SMILES: OB(O)C1=CC=CC(\C=C\C(O)=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results