accessibility menu, dialog, popup

UserName

CAS RN 213749-91-8

H3CCH3ONOCH3N

CAS RN 213749-91-8

IUPAC Name: 2-[4-(cyanomethyl)-5-[(2-ethylhexyl)oxy]-2-methoxyphenyl]acetonitrile
Synonyms: 2-[4-(cyanomethyl)-5-[(2-ethylhexyl)oxy]-2-methoxyphenyl]acetonitrile
Molecular Weight (g/mol): 314.43
Molecular Formula: C19H26N2O2
InChi Key: YGAQISIJKKIHRN-UHFFFAOYNA-N
SMILES: CCCCC(CC)COC1=CC(CC#N)=C(OC)C=C1CC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results