CAS RN 2097002-61-2
CAS RN 2097002-61-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Selitrectinib, MedChemExpress
MedChemExpress Selitrectinib (LOXO-195) is a next-generation TRK kinase inhibitor, with IC50s of 0.6 nM and TRKA and TRKC, respectively.