accessibility menu, dialog, popup

UserName

CAS RN 2096341-92-1

BSOOOHOOH

CAS RN 2096341-92-1

IUPAC Name: {3-[(benzyloxy)sulfonyl]phenyl}boronic acid
Synonyms: 3-(benzyloxysulfonyl)phenylboronic acid
Molecular Weight (g/mol): 292.11
Molecular Formula: C13H13BO5S
InChi Key: UQSAQPVURLUCBP-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC=C1)S(=O)(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results