accessibility menu, dialog, popup

UserName

CAS RN 208516-15-8

BSOONHH3CCH3H3CHOOH

CAS RN 208516-15-8

IUPAC Name: [4-(tert-butylsulfamoyl)phenyl]boronic acid
Synonyms: 4-(tert-butylsulfamoyl)phenylboronic acid
Molecular Weight (g/mol): 257.11
Molecular Formula: C10H16BNO4S
InChi Key: BBTSLOMTYZIGGC-UHFFFAOYSA-N
SMILES: CC(C)(C)NS(=O)(=O)C1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results