accessibility menu, dialog, popup

UserName

CAS RN 199447-10-4

NH2NHOOO(E)

CAS RN 199447-10-4

IUPAC Name: 3-[(E)-N'-hydroxycarbamimidoyl]benzoate
Synonyms: 3-[(E)-N'-hydroxycarbamimidoyl]benzoate
Molecular Weight (g/mol): 179.16
Molecular Formula: C8H7N2O3
InChi Key: LJCLPFBIVUPTAR-UHFFFAOYSA-M
SMILES: N\C(=N\O)C1=CC=CC(=C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results