accessibility menu, dialog, popup

UserName

CAS RN 19781-26-1

H3CCH3OHCH3

CAS RN 19781-26-1

IUPAC Name: 4-ethyloctan-3-ol
Synonyms: 4-ethyloctan-3-ol
Molecular Weight (g/mol): 158.29
Molecular Formula: C10H22O
InChi Key: VXSHGJKAEKIELL-UHFFFAOYNA-N
SMILES: CCCCC(CC)C(O)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results