CAS RN 19666-30-9
CAS RN 19666-30-9
IUPAC Name:
5-tert-butyl-3-[2,4-dichloro-5-(propan-2-yloxy)phenyl]-2,3-dihydro-1,3,4-oxadiazol-2-one
Synonyms:
oxadiazon
Molecular Weight (g/mol):
345.22
Molecular Formula:
C15H18Cl2N2O3
InChi Key:
CHNUNORXWHYHNE-UHFFFAOYSA-N
SMILES:
CC(C)OC1=CC(N2N=C(OC2=O)C(C)(C)C)=C(Cl)C=C1Cl