CAS RN 19646-07-2
CAS RN 19646-07-2
IUPAC Name:
2,4-dichloro-5-methoxypyrimidine
Synonyms:
2,4-dichloro-5-methoxypyrimidine
Molecular Weight (g/mol):
179
Molecular Formula:
C5H4Cl2N2O
InChi Key:
ZTHHRSBDBPCCMZ-UHFFFAOYSA-N
SMILES:
COC1=CN=C(Cl)N=C1Cl
1
–
1
of
1
results
1
2,4-Dichloro-5-methoxypyrimidine, 97%, Thermo Scientific Chemicals
CAS: 19646-07-2 Molecular Formula: C5H4Cl2N2O Molecular Weight (g/mol): 179.00 MDL Number: MFCD02685670 InChI Key: ZTHHRSBDBPCCMZ-UHFFFAOYSA-N Synonym: 2,4-dichloro-5-methoxy-pyrimidine,2,4-dichloro-5-methoxy pyrimidine,pyrimidine, 2,4-dichloro-5-methoxy,2,4-dichloropyrimidin-5-yl methyl ether,5-methoxy-2,4-dichloro pyrimidine,pubchem7076,ksc490s5l,2,4 dichloro-5-methoxypyrimidine,2,4-dichloro-5-methoxy-pyrimidin,2,6-dichloro-5-methoxy pyrimidine PubChem CID: 601401 IUPAC Name: 2,4-dichloro-5-methoxypyrimidine SMILES: COC1=CN=C(Cl)N=C1Cl