accessibility menu, dialog, popup

UserName

CAS RN 19617-44-8

H3COOHNOOCH3

CAS RN 19617-44-8

IUPAC Name: ethyl N-(ethoxycarbonyl)carbamate
Synonyms: ethyl N-(ethoxycarbonyl)carbamate
Molecular Weight (g/mol): 161.16
Molecular Formula: C6H11NO4
InChi Key: PQVSTLUFSYVLTO-UHFFFAOYSA-N
SMILES: CCOC(=O)NC(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)