accessibility menu, dialog, popup

UserName

CAS RN 1949720-50-6

N(R)(R)N(R)N(R)OOPPO(R)(R)(S)(S)NNOOOOOOOOHHONH2NH2HOOHNaNa

CAS RN 1949720-50-6

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)