accessibility menu, dialog, popup

UserName

CAS RN 192863-36-8

BOH3CFFFK

CAS RN 192863-36-8

IUPAC Name: potassium trifluoro(4-methoxyphenyl)boranuide
Synonyms: potassium trifluoro(4-methoxyphenyl)boranuide
Molecular Weight (g/mol): 214.04
Molecular Formula: C7H7BF3KO
InChi Key: XNYMCUFKZHRYID-UHFFFAOYSA-N
SMILES: [K+].COC1=CC=C(C=C1)[B-](F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results