accessibility menu, dialog, popup

UserName

CAS RN 18917-91-4

H3COHOOAlOOCH3HOOOCH3OH

CAS RN 18917-91-4

IUPAC Name: bis[(2-hydroxypropanoyl)oxy]alumanyl 2-hydroxypropanoate
Synonyms: aluminum lactate aluminium trilactate tris(lactato)aluminium
Molecular Weight (g/mol): 294.19
Molecular Formula: C9H15AlO9
InChi Key: VXYADVIJALMOEQ-UHFFFAOYNA-K
SMILES: CC(O)C(=O)O[Al](OC(=O)C(C)O)OC(=O)C(C)O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results