CAS RN 1879-77-2
CAS RN 1879-77-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Doxybetasol, TRC
CAS: 1879-77-2 Molecular Formula: C22 H29 F O4 Molecular Weight (g/mol): 376.46 Synonym: (11β,16β)-9-Fluoro-11,17-dihydroxy-16-methyl-pregna-1,4-diene-3,20-dione,Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,17-dihydroxy-16β-methyl- (7CI,8CI),21-Deoxybetamethasone,9-Fluoro-11β,17-dihydroxy-16β-methylpregna-1,4-diene-3,20-dione,Doxibetasol,Doxybetasol,Betamethasone Valerate Imp. B (EP) IUPAC Name: (8S,9R,10S,11S,13S,14S,16S,17R)-17-acetyl-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one SMILES: C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)C