accessibility menu, dialog, popup

UserName

CAS RN 18683-91-5

NH2BrBrNHHO

CAS RN 18683-91-5

IUPAC Name: 4-{[(2-amino-3,5-dibromophenyl)methyl]amino}cyclohexan-1-ol
Synonyms: ambroβ
Molecular Weight (g/mol): 378.11
Molecular Formula: C13H18Br2N2O
InChi Key: JBDGDEWWOUBZPM-UHFFFAOYSA-N
SMILES: NC1=C(Br)C=C(Br)C=C1CNC1CCC(O)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results