accessibility menu, dialog, popup

UserName

CAS RN 1857-57-4

H3COOHOBr

CAS RN 1857-57-4

IUPAC Name: 3-(3-bromo-4-methoxyphenyl)propanoic acid
Synonyms: 3-(3-bromo-4-methoxyphenyl)propanoic acid
Molecular Weight (g/mol): 259.1
Molecular Formula: C10H11BrO3
InChi Key: MLXVZRVWVCOUBQ-UHFFFAOYSA-N
SMILES: COC1=C(Br)C=C(CCC(O)=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results