accessibility menu, dialog, popup

UserName

CAS RN 18529-01-6

H3CClFN

CAS RN 18529-01-6

IUPAC Name: 4-chloro-6-fluoro-2-methylquinoline
Synonyms: 4-chloro-6-fluoro-2-methylquinoline
Molecular Weight (g/mol): 195.62
Molecular Formula: C10H7ClFN
InChi Key: GMMVVJJBQBYMNZ-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=C2C=C(F)C=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results