CAS RN 183204-72-0
CAS RN 183204-72-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tipiracil hydrochloride, MedChemExpress
MedChemExpress Tipiracil (hydrochloride) is a thymidine phosphorylase inhibitor (TPI), used for cancer research.