CAS RN 17977-76-3
CAS RN 17977-76-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Chloro-N-[4-chloro-2-[(hydroxyimino)phenylmethyl]phenyl]acetamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.