accessibility menu, dialog, popup

UserName

CAS RN 177834-92-3

BrCH3N(R)HNSOOH

CAS RN 177834-92-3

IUPAC Name: hydrogen 5-[2-(benzenesulfonyl)ethyl]-3-{[(2R)-1-methylpyrrolidin-2-yl]methyl}-1H-indole bromide
Synonyms: hydrogen eletriptan bromide
Molecular Weight (g/mol): 463.43
Molecular Formula: C22H27BrN2O2S
InChi Key: UTINOWOSWSPFLJ-FSRHSHDFSA-N
SMILES: [H+].[Br-].CN1CCC[C@@H]1CC1=CNC2=CC=C(CCS(=O)(=O)C3=CC=CC=C3)C=C12
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results