CAS RN 17403-07-5
CAS RN 17403-07-5
IUPAC Name:
3-(1-methylpiperidin-4-yl)-1H-indole
Synonyms:
3-(1-methylpiperidin-4-yl)-1H-indole
Molecular Weight (g/mol):
214.31
Molecular Formula:
C14H18N2
InChi Key:
KYSCKYJNMTUJPA-UHFFFAOYSA-N
SMILES:
CN1CCC(CC1)C1=CNC2=CC=CC=C12
1
–
2
of
2
results
1
3-(1-Methyl-4-piperidinyl)indole, 97%
CAS: 17403-07-5 Molecular Formula: C14H18N2 Molecular Weight (g/mol): 214.31 MDL Number: MFCD07776765 InChI Key: KYSCKYJNMTUJPA-UHFFFAOYSA-N PubChem CID: 11206672 IUPAC Name: 3-(1-methylpiperidin-4-yl)-1H-indole SMILES: CN1CCC(CC1)C1=CNC2=CC=CC=C12
2
3-(1-Methylpiperidin-4-yl)-1H-indole, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.