accessibility menu, dialog, popup

UserName

CAS RN 17249-80-8

ClS

CAS RN 17249-80-8

IUPAC Name: 3-chlorothiophene
Synonyms: 3-chlorothiophene
Molecular Weight (g/mol): 118.58
Molecular Formula: C4H3ClS
InChi Key: QUBJDMPBDURTJT-UHFFFAOYSA-N
SMILES: ClC1=CSC=C1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)