CAS RN 17066-67-0
CAS RN 17066-67-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Beta-Selinene, (~90%), TRC
CAS: 17066-67-0 Molecular Formula: C15H24 Molecular Weight (g/mol): 204.35 Synonym: [4aR-(4aalpha,7alpha,8abeta)]-Decahydro-4a-methyl-1-methylene-7-(1-methylethenyl)-naphthalene,Eudesma-4(14),11-diene,Selina-4(14),11-diene,beta-Eudesmene,(4aR,7R,8aS)-Decahydro-4a-methyl-1-methylene-7-(1-methylethenyl)naphthalene;(+)-beta-Selinene,Decahydro-4a-methyl-1-methylene-7-(1-methylethenyl)-[4aR-(4aalpha,7alpha,8abeta)]-naphthalene; IUPAC Name: (3R,4aS,8aR)-8a-methyl-5-methylidene-3-prop-1-en-2-yl-1,2,3,4,4a,6,7,8-octahydronaphthalene SMILES: CC(=C)[C@@H]1CC[C@@]2(C)CCCC(=C)[C@@H]2C1