accessibility menu, dialog, popup

UserName

CAS RN 170434-06-7

H3CNNCl

CAS RN 170434-06-7

IUPAC Name: 2-chloro-5-decylpyrimidine
Synonyms: 2-chloro-5-decylpyrimidine
Molecular Weight (g/mol): 254.8
Molecular Formula: C14H23ClN2
InChi Key: LJRSECCCAXFYFU-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC1=CN=C(Cl)N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results