accessibility menu, dialog, popup

UserName

CAS RN 16898-52-5

NHNH

CAS RN 16898-52-5

IUPAC Name: 4-[3-(piperidin-4-yl)propyl]piperidine
Synonyms: 4-[3-(piperidin-4-yl)propyl]piperidine
Molecular Weight (g/mol): 210.37
Molecular Formula: C13H26N2
InChi Key: OXEZLYIDQPBCBB-UHFFFAOYSA-N
SMILES: C(CC1CCNCC1)CC1CCNCC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results