accessibility menu, dialog, popup

UserName

CAS RN 16849-88-0

CH3NH3CNN

CAS RN 16849-88-0

IUPAC Name: 2-[(dimethylamino)methylidene]propanedinitrile
Synonyms: 2-[(dimethylamino)methylidene]propanedinitrile
Molecular Weight (g/mol): 121.14
Molecular Formula: C6H7N3
InChi Key: LBUDLOYYNHQKQI-UHFFFAOYSA-N
SMILES: CN(C)C=C(C#N)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results