accessibility menu, dialog, popup

UserName

CAS RN 16801-63-1

ONHOO

CAS RN 16801-63-1

IUPAC Name: benzyl N-(4-oxocyclohexyl)carbamate
Synonyms: benzyl N-(4-oxocyclohexyl)carbamate
Molecular Weight (g/mol): 247.29
Molecular Formula: C14H17NO3
InChi Key: VHXBIBFCJISKFA-UHFFFAOYSA-N
SMILES: O=C(NC1CCC(=O)CC1)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results