accessibility menu, dialog, popup

UserName

CAS RN 16801-62-0

HNOOHO

CAS RN 16801-62-0

IUPAC Name: benzyl N-(4-hydroxycyclohexyl)carbamate
Synonyms: benzyl N-(4-hydroxycyclohexyl)carbamate
Molecular Weight (g/mol): 249.31
Molecular Formula: C14H19NO3
InChi Key: JNPBKQOVSMBPGE-UHFFFAOYSA-N
SMILES: OC1CCC(CC1)NC(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results