CAS RN 16640-68-9
CAS RN 16640-68-9
IUPAC Name:
2-(triphenyl-λ⁵-phosphanylidene)acetonitrile
Synonyms:
2-(triphenyl-λ⁵-phosphanylidene)acetonitrile
Molecular Weight (g/mol):
301.33
Molecular Formula:
C20H16NP
InChi Key:
APISVOVOJVZIBA-UHFFFAOYSA-N
SMILES:
N#CC=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1