CAS RN 1646570-98-0
CAS RN 1646570-98-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
4-Bromo-1-propan-2-ylpyridin-2-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Bromo-1-propan-2-ylpyridin-2-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Bromo-1-propan-2-ylpyridin-2-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.