CAS RN 1643131-93-4
CAS RN 1643131-93-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,5-Ethyl Acetate Febuxostat, TRC
CAS: 1643131-93-4 Molecular Formula: C20 H25 N O5 S Molecular Weight (g/mol): 391.48 Synonym: Febuxostat Impurity,Febuxostat DEE Impurity,Ethyl 2-(3-(Ethoxycarbonyl)-4-isobutoxyphenyl)-4-methylthiazole-5-carboxylate,2-[3-(Ethoxycarbonyl)-4-(2-methylpropoxy)phenyl]-4-methyl-5-thiazolecarboxylic Acid Ethyl Ester IUPAC Name: ethyl 2-(3-ethoxycarbonyl-4-isobutoxy-phenyl)-4-methyl-thiazole-5-carboxylate SMILES: CCOC(=O)c1cc(ccc1OCC(C)C)c2nc(C)c(s2)C(=O)OCC