CAS RN 1609651-90-2
CAS RN 1609651-90-2
IUPAC Name:
3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-N-(3,6,9,12-tetraoxapentadec-14-yn-1-yl)propanamide
Synonyms:
3-(2,5-dioxopyrrol-1-yl)-N-(3,6,9,12-tetraoxapentadec-14-yn-1-yl)propanamide
Molecular Weight (g/mol):
382.41
Molecular Formula:
C18H26N2O7
InChi Key:
GHEBECZRHJXPTL-UHFFFAOYSA-N
SMILES:
O=C(CCN1C(=O)C=CC1=O)NCCOCCOCCOCCOCC#C
1
–
1
of
1
results
1
Acetylene-PEG4-maleimide
CAS: 1609651-90-2 Molecular Formula: C18H26N2O7 Molecular Weight (g/mol): 382.41 MDL Number: MFCD22380740 InChI Key: GHEBECZRHJXPTL-UHFFFAOYSA-N Synonym: acetylene-peg4-maleimide,propargyl-peg4-maleimide,alkyne-peg4-maleimide hplc PubChem CID: 91757777